Merck
  • Production of Jet Fuel-Range Hydrocarbons from Hydrodeoxygenation of Lignin over Super Lewis Acid Combined with Metal Catalysts.
Production of Jet Fuel-Range Hydrocarbons from Hydrodeoxygenation of Lignin over Super Lewis Acid Combined with Metal Catalysts.
ChemSusChem (2017-11-15)
Hongliang Wang, Huamin Wang, Eric Kuhn, Melvin P Tucker, Bin Yang
ABSTRACT
Super Lewis acids containing the triflate anion [e.g., Hf(OTf)
Sigma-Aldrich
Indium(III) trifluoromethanesulfonate
Sigma-Aldrich
Benzyl phenyl ether, 98%