- HID-1 is a novel player in the regulation of neuropeptide sorting.
HID-1 is a novel player in the regulation of neuropeptide sorting.
The Biochemical journal (2011-01-22)
Yong Yu, Lifen Wang, Yaming Jiu, Yi Zhan, Lin Liu, Zhiping Xia, Eli Song, Pingyong Xu, Tao Xu
PMID21250940
ABSTRACT
Peptide hormones and neuropeptides are packaged and stored in a specialized intracellular organelle called the dense core vesicle. It remains elusive how peptide cargoes are correctly sorted. In the present study, we show that a highly conserved Golgi-localized protein named HID-1 acts to prevent mis-sorting of peptide cargoes to lysosomes for degradation via a PtdIns3P-dependent trafficking pathway. Epistasis analysis suggests that rab-2 is epistatic to hid-1.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Anti-TGN46 antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution