Merck
Improvement in xylooligosaccharides production by knockout of the
3 Biotech (2017-12-28)
Chuannan Long, Jingjing Cui, Hailong Li, Jian Liu, Lihui Gan, Bin Zeng, Minnan Long
ABSTRACT
The goal of this study was to enhance the production of xylooligosaccharides (XOs) and reduce the production of xylose. We investigated β-xylosidases, which were key enzymes in the hydrolysis of xylan into xylose, in
Sigma-Aldrich
Xylobiose, ≥90% (HPLC)