Merck
  • Characterization of Toxoplasma DegP, a rhoptry serine protease crucial for lethal infection in mice.
Characterization of Toxoplasma DegP, a rhoptry serine protease crucial for lethal infection in mice.
PloS one (2017-12-16)
Gaelle Lentini, Hiba El Hajj, Julien Papoin, Gamou Fall, Alexander W Pfaff, Nadim Tawil, Catherine Braun-Breton, Maryse Lebrun
ABSTRACT
During the infection process, Apicomplexa discharge their secretory organelles called micronemes, rhoptries and dense granules to sustain host cell invasion, intracellular replication and to modulate host cell pathways and immune responses. Herein, we describe the Toxoplasma gondii Deg-like serine protein (TgDegP), a rhoptry protein homologous to High temperature requirement A (HtrA) or Deg-like family of serine proteases. TgDegP undergoes processing in both types I and II strains as most of the rhoptries proteins. We show that genetic disruption of the degP gene does not impact the parasite lytic cycle in vitro but affects virulence in mice. While in a type I strain DegPI appears dispensable for the establishment of an infection, removal of DegPII in a type II strain dramatically impairs the virulence. Finally, we show that KO-DegPII parasites kill immunodeficient mice as efficiently as the wild-type strain indicating that the protease might be involved in the complex crosstalk that the parasite engaged with the host immune response. Thus, this study unravels a novel rhoptry protein in T. gondii important for the establishment of lethal infection.
Sigma-Aldrich
ML277, ≥98% (HPLC)