Merck
Structural insights into the potency of SK channel positive modulators.
Scientific reports (2017-12-08)
Young-Woo Nam, Razan Orfali, Tingting Liu, Kunqian Yu, Meng Cui, Heike Wulff, Miao Zhang
ABSTRACT
Small-conductance Ca
Sigma-Aldrich
1-EBIO, ≥98% (HPLC)
Pricing and availability is not currently available.