- Biostimulant Effects of Seed-Applied Sedaxane Fungicide: Morphological and Physiological Changes in Maize Seedlings.
Biostimulant Effects of Seed-Applied Sedaxane Fungicide: Morphological and Physiological Changes in Maize Seedlings.
Frontiers in plant science (2017-12-23)
Cristian Dal Cortivo, Giovanni Battista Conselvan, Paolo Carletti, Giuseppe Barion, Luca Sella, Teofilo Vamerali
PMID29270181
ABSTRACT
Most crops are routinely protected against seed-born and soil-borne fungal pathogens through seed-applied fungicides. The recently released succinate dehydrogenase inhibitor (SDHI), sedaxane
MATERIALS