Merck
Ambient-Stable Bis-Azoaromatic-Centered Diradical [(L
Inorganic chemistry (2017-10-14)
Sima Roy, Shuvam Pramanik, Sarat Chandra Patra, Basab Adhikari, Abhishake Mondal, Sanjib Ganguly, Kausikisankar Pramanik
ABSTRACT
Bis-azoaromatic electron traps, viz. 2-(2-pyridylazo)azoarene 1, have been synthesized by colligating electron-deficient pyridine and azoarene moieties, and they act as apposite proradical templates for the formation of stable open-shell diradical complexes [(1
Sigma-Aldrich
Rhodium(III) chloride, 98%