Merck
  • α1-Blockers in Men with Lower Urinary Tract Symptoms Suggestive of Benign Prostatic Obstruction: Is Silodosin Different?
α1-Blockers in Men with Lower Urinary Tract Symptoms Suggestive of Benign Prostatic Obstruction: Is Silodosin Different?
Advances in therapy (2016-11-05)
Claus G Roehrborn, Francisco Cruz, Ferdinando Fusco
ABSTRACT
Available α1-blockers (ABs) have different profiles of receptor selectivity. Silodosin exhibits the highest selectivity for the α
Sigma-Aldrich
Silodosin, ≥98% (HPLC)