Merck
  • Spontaneous vesicle formation and vesicle-to-micelle transition of sodium 2-ketooctanate in water.
Spontaneous vesicle formation and vesicle-to-micelle transition of sodium 2-ketooctanate in water.
Journal of colloid and interface science (2017-09-16)
Huifang Xu, Na Du, Yawen Song, Shue Song, Wanguo Hou
ABSTRACT
Single-tailed short-chain alkyl keto-acids/salts, a class of fatty acid/salt derivatives, such as sodium 2-ketooctanate (KOCOONa), are a kind of weakly acid/salt type amphiphiles and plausible prebiotic molecules, and the current understanding of their aggregation behavior in aqueous solutions is still limited. Herein, the aggregation behavior of KOCOONa in aqueous solution was studied by changing its concentration (C), using equilibrium surface tension, conductivity, and fluorescence measurements. The aggregates formed were characterized using freeze-fracture and cryogenic transmission electron microscopy, dynamic light scattering, atomic force microscopy, and confocal laser scanning microscopy. A concentration-driven stepwise aggregation was identified in the KOCOONa solution. Vesicles can spontaneously form from the single-component aqueous solution, with a critical vesicle concentration (CVC) of ∼15mM, which is obviously lower than that of octanoic acid/salt (120-200mM). With increasing C, a vesicle-to-micelle transition can occur, showing a critical micelle concentration (CMC) of ∼80mM. In addition, the membrane permeability of the KOCOONa vesicles was examined using small-size Calcein and large-size FITC-BSA as fluorescence probes, showing a size-selective permeability, similar to short-chain (C8-C11) fatty acid vesicles. For the first time, the aggregation behavior of single-tailed keto-acid salt surfactant is reported.
Sigma-Aldrich
2-Oxooctanoic acid, ≥99.0% (T)
Sigma-Aldrich
Calcein, Used for the fluorometric determination of calcium and EDTA titration of calcium in the presence of magnesium.