Merck
  • Proteomic Data in Morphine Addiction Versus Real Protein Activity: Metabolic Enzymes.
Proteomic Data in Morphine Addiction Versus Real Protein Activity: Metabolic Enzymes.
Journal of cellular biochemistry (2017-04-22)
Anna Antolak, Anna Bodzoń-Kułakowska, Ewa Cetnarska, Monika Pietruszka, Marta Marszałek-Grabska, Jolanta Kotlińska, Piotr Suder
ABSTRACT
Drug dependence is an escalating problem worldwide and many efforts are being made to understand the molecular basis of addiction. The morphine model is widely used in these investigations. To date, at least 29 studies exploring the influence of morphine on mammals' proteomes have been published. Among various proteins indicated as up- or down-regulated, the expression changes of enzymes engaged in energy metabolism pathways have often been confirmed. To verify whether proteomics-indicated alterations in enzyme levels reflect changes in their activity, four enzymes: PK, MDH, Complex I, and Complex V were investigated in morphine addiction and abstinence models. After analyses of the rat brain mitochondria fraction in the model of morphine dependence, we found that one of the investigated enzymes (pyruvate kinase) showed statistically significant differences observed between morphine, control, and abstinence groups. J. Cell. Biochem. 118: 4323-4330, 2017. © 2017 Wiley Periodicals, Inc.
Sigma-Aldrich
Pyruvate Kinase Activity Assay Kit, sufficient for 100 colorimetric or fluorometric tests
Sigma-Aldrich
Malate Dehydrogenase Assay Kit, Sufficient for 100 Colorimetric tests
Supelco
Bradford Reagent, for 0.1-1.4 mg/ml protein