Merck
  • CYP17A1-independent production of the neurosteroid-derived 5α-pregnan-3β,6α-diol-20-one in androgen-responsive prostate cancer cell lines under serum starvation and inhibition by Abiraterone.
CYP17A1-independent production of the neurosteroid-derived 5α-pregnan-3β,6α-diol-20-one in androgen-responsive prostate cancer cell lines under serum starvation and inhibition by Abiraterone.
The Journal of steroid biochemistry and molecular biology (2017-09-12)
Antonio G Gomes de Mello Martins, Giuseppe Allegretta, Gerhard Unteregger, Jörg Haupenthal, Jens Eberhard, Michael Hoffmann, Jill A van der Zee, Kerstin Junker, Michael Stöckle, Rolf Müller, Rolf W Hartmann, Carsten-H Ohlmann
ABSTRACT
CYP17A1-independent intratumoral steroid hormone synthesis is regarded as one possible explanation for resistance to treatment with the CYP17-inhibitor Abiraterone (Abi). The aim of our study was therefore to investigate the steroid metabolism of prostate cancer cells under serum starvation and the effects of Abi treatment. We assessed steroid metabolism in a panel of prostate cancer cells under serum starvation by radioactivity detector-coupled HPLC and HPLC-ESI-ToF-mass spectrometry after treatment with pregnenolone, progesterone and allopregnanolone. We further evaluated the effects of Abi on steroid metabolism of testosterone, dihydrotestosterone (DHT) and dehydroepiandrosterone (DHEA) by enzyme immunoassays (EIAs). Androgen-responsive cell lines metabolized pregnenolone primarily to mitogenic steroid 5α-pregnan-3β,6α-diol-20-one under serum starvation. Co-administration of Abi lead to detectable concentrations of the Abi metabolite Δ
Sigma-Aldrich
Pregnenolone-20,21-13C2-16,16-d2, ≥98 atom %, ≥98% (CP)
Sigma-Aldrich
5-Pregnen-3β-ol-20-one, ≥98%
Sigma-Aldrich
5α-Pregnan-3α-ol-20-one, solid