Merck
The tumor suppressor RASSF1A induces the YAP1 target gene
Oncotarget (2017-11-29)
Adriana P Jiménez, Annalena Traum, Thomas Boettger, Holger Hackstein, Antje M Richter, Reinhard H Dammann
ABSTRACT
The Hippo pathway regulates organ size, growth and comprises several tumor related factors, including the oncoprotein YAP1 and the tumor suppressor RASSF1A.
Sigma-Aldrich
MISSION® esiRNA, targeting human ANKRD1