Merck
  • Exogenous proline stimulates type I collagen and HIF-1α expression and the process is attenuated by glutamine in human skin fibroblasts.
Exogenous proline stimulates type I collagen and HIF-1α expression and the process is attenuated by glutamine in human skin fibroblasts.
Molecular and cellular biochemistry (2017-05-21)
Lukasz Szoka, Ewa Karna, Kornelia Hlebowicz-Sarat, Jacek Karaszewski, Jerzy A Palka
ABSTRACT
Abundance of proline (Pro) in collagen molecule led us to investigate whether Pro supply affects collagen biosynthesis in human skin fibroblasts. Treatment of the cells with milimolar concentrations (5 and 10 mM) of Pro for 24 and 48 h contributed to increase in α
Sigma-Aldrich
Anti-Goat IgG (whole molecule)–Peroxidase antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-MAP Kinase, Activated (Diphosphorylated ERK-1&2) antibody produced in mouse, clone MAPK-YT, ascites fluid
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-IκBα antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
L-4-Thiazolidinecarboxylic acid, 98%
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution