Merck
  • Autophagy maintains the integrity of endothelial barrier in LPS-induced lung injury.
Autophagy maintains the integrity of endothelial barrier in LPS-induced lung injury.
Journal of cellular physiology (2017-03-23)
Dan Zhang, Jian Zhou, Le Chi Ye, Jing Li, Zhenzhou Wu, Yuping Li, Chichi Li
ABSTRACT
Understanding the role and underlying regulation mechanism of autophagy in lipopolysaccharide-induced lung injury (LPS-LI) may provide potentially new pharmacological targets for treatment of acute lung injury. The aim of this study was to investigate the functional significance of autophagy in LPS-LI. The autophagy of human pulmonary microvascular endothelial cells (HPMVECs) and mice was inhibited before they were challenged with LPS. In vitro, permeability, vitality, and the LDH release rate of the cells were detected, the zonula occluden-1 (ZO-1) expression and the stress fiber formation were determined. In vivo, the lung injury was assessed. We found LPS caused high permeability and increased lactate dehydrogenase (LDH) release rate, lowered viability of the cells, inhibited the ZO-1 expression and induced stress fiber formation, these effects were further aggravated by prohibiting the level of autophagy. Consistently, in in vivo experiments, LPS-induced serious lung injury, which was reflected as edema, leukocyte infiltration and hemorrhage in lung tissue, and the high concentration of pro-inflammation cytokines tumor necrosis factor (TNF)-α and interleukin (IL)-1β in bronchoalveolar lavage fluid (BALF). Inhibiting autophagy further exacerbated LPS-LI. It appears that autophagy played a protective role in LPS-LI in part through restricting the injury of lung microvascular barrier.
Sigma-Aldrich
Chloroquine diphosphate salt, powder or crystals, 98.5-101.0% (EP)
Pricing and availability is not currently available.
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O111:B4, purified by phenol extraction
Pricing and availability is not currently available.
Sigma-Aldrich
FITC-CM-Dextran, average mol wt 40,000
Pricing and availability is not currently available.