Merck
  • Impact of Endothelial Lipase on Cholesterol Efflux Capacity of Serum and High-density Lipoprotein.
Impact of Endothelial Lipase on Cholesterol Efflux Capacity of Serum and High-density Lipoprotein.
Scientific reports (2017-10-04)
Irene Schilcher, Sabine Kern, Andelko Hrzenjak, Thomas O Eichmann, Tatjana Stojakovic, Hubert Scharnagl, Madalina Duta-Mare, Dagmar Kratky, Gunther Marsche, Saša Frank
ABSTRACT
Endothelial lipase (EL) is a potent modulator of the structural and functional properties of HDL. Impact of EL on cholesterol efflux capacity (CEC) of serum and isolated HDL is not well understood and apparently contradictory data were published. Here, we systematically examined the impact of EL on composition and CEC of serum and isolated HDL, in vitro and in vivo, using EL-overexpressing cells and EL-overexpressing mice. CEC was examined in a validated assay using
Sigma-Aldrich
Sandoz 58-035, >98% (HPLC), powder