- Phosphine inhibits transcription of the catalase gene through the DRE/DREF system in Drosophila melanogaster.
Phosphine inhibits transcription of the catalase gene through the DRE/DREF system in Drosophila melanogaster.
Scientific reports (2017-10-12)
Tao Liu, Li Li, Baishu Li, Guoping Zhan
PMID29018235
ABSTRACT
Phosphine (PH
Product Description
Mininert® Valve, screw thread, for use with 24/400 mm thread, pkg of 12 ea
Sigma-Aldrich
Wheaton narrow mouth bottles with caps, capacity 1 oz (30 mL), amber, white polypropylene cap, PTFE flat liner