- n-Alcohol Length Governs Shift in L
n-Alcohol Length Governs Shift in L
Biophysical journal (2017-08-13)
Caitlin E Cornell, Nicola L C McCarthy, Kandice R Levental, Ilya Levental, Nicholas J Brooks, Sarah L Keller
PMID28801104
ABSTRACT
A persistent challenge in membrane biophysics has been to quantitatively predict how membrane physical properties change upon addition of new amphiphiles (e.g., lipids, alcohols, peptides, or proteins) in order to assess whether the changes are large enough to plausibly result in biological ramifications. Because of their roles as general anesthetics, n-alcohols are perhaps the best-studied amphiphiles of this class. When n-alcohols are added to model and cell membranes, changes in membrane parameters tend to be modest. One striking exception is found in the large decrease in liquid-liquid miscibility transition temperatures (T
MATERIALS