- Synthesis and Preliminary Studies of a Novel Negative Allosteric Modulator, 7-((2,5-Dioxopyrrolidin-1-yl)methyl)-4-(2-fluoro-4-[
Synthesis and Preliminary Studies of a Novel Negative Allosteric Modulator, 7-((2,5-Dioxopyrrolidin-1-yl)methyl)-4-(2-fluoro-4-[
ACS chemical neuroscience (2017-06-02)
Xiaofei Zhang, Katsushi Kumata, Tomoteru Yamasaki, Ran Cheng, Akiko Hatori, Longle Ma, Yiding Zhang, Lin Xie, Lu Wang, Hye Jin Kang, Douglas J Sheffler, Nicholas D P Cosford, Ming-Rong Zhang, Steven H Liang
PMID28565908
ABSTRACT
Metabotropic glutamate 2 receptors (mGlu