- Post-translational modification of HINT1 mediates activation of MITF transcriptional activity in human melanoma cells.
Post-translational modification of HINT1 mediates activation of MITF transcriptional activity in human melanoma cells.
Microphthalmia transcription factor (MITF) is a basic helix-loop-helix leucine zipper (bHLH-Zip) DNA-binding protein. This transcription factor plays a crucial role in the physiological and pathological functions of distinct cell types. MITF transcriptional activity is inhibited by the histidine triad nucleotide-binding protein 1 (HINT1) through direct binding. We previously reported that this association is disrupted by the binding of the second messenger Ap