Merck
  • Gemcitabine-camptothecin conjugates: a hybrid prodrug for controlled drug release and synergistic therapeutics.
Gemcitabine-camptothecin conjugates: a hybrid prodrug for controlled drug release and synergistic therapeutics.
Biomaterials science (2017-07-07)
Meili Hou, Peng Xue, Yong-E Gao, Xiaoqian Ma, Shuang Bai, Yuejun Kang, Zhigang Xu
ABSTRACT
Drug self-delivery systems represent an important approach to enhance the therapeutic efficacy for cancer therapy. We report the design, synthesis and characterization of a new amphiphilic small molecule prodrug based on two types of anticancer drugs, the hydrophilic gemcitabine and hydrophobic camptothecin, linked by a disulfide bond and abbreviated as GT-CPT. The obtained amphiphilic prodrug conjugates self-assembled into nanoparticles in water and showed strong micellar stability and excellent blood compatibility in vivo. The GT-CPT prodrug conjugates could realize precise drug loading as high as ∼75 wt% demonstrating a carrier-free model for efficient drug delivery. Furthermore, the reduction-responsive disulfide bond enabled controlled drug release in the presence of tumour-specific microenvironment. It was found that each of these hybrid drug components (CPT and GT) not only showed enhanced cytotoxicity individually but also exhibited a prominent synergistic effect on HeLa and MCF-7 cancer cells. This study demonstrated the promising potential of this stimuli-responsive hybrid prodrug conjugate for highly efficient co-delivery of multiple anticancer chemotherapeutics, which could inspire further applications using such hybrid prodrug conjugates for combination cancer chemotherapy.
Sigma-Aldrich
Triphosgene, reagent grade, 98%
Sigma-Aldrich
Pyrene, sublimed grade, 99%