- Development of a monoclonal anti-ADAMTS-5 antibody that specifically blocks the interaction with LRP1.
Development of a monoclonal anti-ADAMTS-5 antibody that specifically blocks the interaction with LRP1.
mAbs (2017-03-18)
Salvatore Santamaria, Oleg Fedorov, John McCafferty, Gillian Murphy, Jayesh Dudhia, Hideaki Nagase, Kazuhiro Yamamoto
PMID28306378
ABSTRACT
The potent aggrecanase ADAMTS-5 is constitutively secreted by chondrocytes, but it is rapidly endocytosed in normal cartilage via the cell surface endocytic receptor LRP1. Therefore it is difficult to detect the total ADAMTS-5 activity produced. In this study, we isolated a monoclonal anti-ADAMTS-5 antibody 1B7 that blocks LRP1-mediated internalization without affecting the aggrecanolytic activity. Addition of 1B7 to cultured human chondrocytes revealed the full aggrecanolytic activity of ADAMTS-5 generated by the cells. 1B7 is a useful tool to estimate the ADAMTS-5 activity and to identify its potential roles in the tissues.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)