Merck
  • Influence of Herbicide Triasulfuron on Soil Microbial Community in an Unamended Soil and a Soil Amended with Organic Residues.
Influence of Herbicide Triasulfuron on Soil Microbial Community in an Unamended Soil and a Soil Amended with Organic Residues.
Frontiers in microbiology (2017-03-25)
Eva Pose-Juan, José M Igual, María J Sánchez-Martín, M S Rodríguez-Cruz
ABSTRACT
The effect of organic amendments and pesticides on a soil microbial community has garnered considerable interest due to the involvement of microorganisms in numerous soil conservation and maintenance reactions. The aim of this work was to assess the influence on a soil microbial community of the simultaneous application of the herbicide triasulfuron at three doses (2, 10, and 50 mg kg
Product Description
Supelco
Triasulfuron, PESTANAL®, analytical standard