Merck
  • Brachytherapy with Intratumoral Injections of Radiometal-Labeled Polymers That Thermoresponsively Self-Aggregate in Tumor Tissues.
Brachytherapy with Intratumoral Injections of Radiometal-Labeled Polymers That Thermoresponsively Self-Aggregate in Tumor Tissues.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine (2017-04-15)
Kohei Sano, Yuko Kanada, Kengo Kanazaki, Ning Ding, Masahiro Ono, Hideo Saji
ABSTRACT
Brachytherapy is a type of radiotherapy wherein titanium capsules containing therapeutic radioisotopes are implanted within tumor tissues, enabling high-dose radioirradiation to tumor tissues around the seeds. Although marked therapeutic effects have been demonstrated, brachytherapy needs a complicated implantation technique under general anesthesia and the seeds could migrate to other organs. The aim of this study was to establish a novel brachytherapy using biocompatible, injectable thermoresponsive polymers (polyoxazoline [POZ]) labeled with
Sigma-Aldrich
2-n-Propyl-2-oxazoline
Pricing and availability is not currently available.