Merck
  • Co-expression of sulphydryl oxidase and protein disulphide isomerase in Escherichia coli allows for production of soluble CRM
Co-expression of sulphydryl oxidase and protein disulphide isomerase in Escherichia coli allows for production of soluble CRM
Journal of applied microbiology (2017-03-10)
R Roth, P van Zyl, T Tsekoa, S Stoychev, S Mamputha, S Buthelezi, M Crampton
ABSTRACT
To investigate the production of soluble cross-reacting material 197 (CRM The use of co-expression of a sulphydryl oxidase (SOX) and protein disulphide isomerase for the production of soluble CRM SOX and protein disulphide isomerase are enzymes involved in the formation of intra-protein disulphide bonds, and can influence the tertiary structure of the protein being produced, resulting in increased solubility due to the correct folding of the protein. Their use enabled the production of soluble untagged CRM Previous literature reports have shown that CRM
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine (TMB) Liquid Substrate System for Membranes, ready to use solution
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine (TMB) Liquid Substrate System for ELISA, peroxidase substrate