- Impacts of Unregulated Novel Brominated Flame Retardants on Human Liver Thyroid Deiodination and Sulfotransferation.
Impacts of Unregulated Novel Brominated Flame Retardants on Human Liver Thyroid Deiodination and Sulfotransferation.
The inhibitory effects of five novel brominated flame retardants, 1,2-bis(2,4,5-tribromophenoxy)ethane (BTBPE), decabromodiphenylethane (DBDPE), 2-ethylhexyl-2,3,4,5-tetrabromobenzoate (EH-TBB), bis(2-ethylhexyl)tetrabromophthalate (BEH-TEBP), and β-tetrabromoethylcyclohexane (β-TBECH), on thyroid hormone deiodinase (DIO) and sulfotransferase (SULT) activity were investigated using human in vitro liver microsomal and cytosolic bioassays. Enzymatic activity was measured by incubating active human liver subcellular fractions with thyroid hormones (T4 and rT3 separately) and measuring changes in thyroid hormone (T4, T3, rT3, and 3,3'-T2) concentrations. Only DBDPE showed inhibition of both outer and inner ring deiodination (O and IRD) of T3 and 3,3'-T2 formation from T4, respectively, with an estimated IC