Merck
Ectopic Expression of PII Induces Stomatal Closure in
Frontiers in plant science (2017-08-10)
Aurora Parlati, Vladimir T Valkov, Enrica D'Apuzzo, Ludovico M Alves, Angelo Petrozza, Stephan Summerer, Alex Costa, Francesco Cellini, Alain Vavasseur, Maurizio Chiurazzi
ABSTRACT
The PII protein in plants has been associated to many different tissue specialized roles concerning the Nitrogen assimilation pathways. We report here the further characterization of
Sigma-Aldrich
DAF-FM DA solution, 5 mM in DMSO, ≥97% (HPLC)
Pricing and availability is not currently available.