Merck
  • Low-dose cadmium exposure induces peribronchiolar fibrosis through site-specific phosphorylation of vimentin.
Low-dose cadmium exposure induces peribronchiolar fibrosis through site-specific phosphorylation of vimentin.
American journal of physiology. Lung cellular and molecular physiology (2017-04-30)
Fu Jun Li, Ranu Surolia, Huashi Li, Zheng Wang, Gang Liu, Rui-Ming Liu, Sergey B Mirov, Mohammad Athar, Victor J Thannickal, Veena B Antony
ABSTRACT
Exposure to cadmium (Cd) has been associated with development of chronic obstructive lung disease (COPD). The mechanisms and signaling pathways whereby Cd causes pathological peribronchiolar fibrosis, airway remodeling, and subsequent airflow obstruction remain unclear. We aimed to evaluate whether low-dose Cd exposure induces vimentin phosphorylation and Yes-associated protein 1 (YAP1) activation leading to peribronchiolar fibrosis and subsequent airway remodeling. Our data demonstrate that Cd induces myofibroblast differentiation and extracellular matrix (ECM) deposition around small (<2 mm in diameter) airways. Upon Cd exposure, α-smooth muscle actin (α-SMA) expression and the production of ECM proteins, including fibronectin and collagen-1, are markedly induced in primary human lung fibroblasts. Cd induces Smad2/3 activation and the translocation of both Smad2/3 and Yes-associated protein 1 (YAP1) into the nucleus. In parallel, Cd induces AKT and cdc2 phosphorylation and downstream vimentin phosphorylation at Ser
Sigma-Aldrich
MISSION® esiRNA, targeting human VIM
Pricing and availability is not currently available.