Merck
  • Application of Mycorrhiza and Soil from a Permaculture System Improved Phosphorus Acquisition in Naranjilla.
Application of Mycorrhiza and Soil from a Permaculture System Improved Phosphorus Acquisition in Naranjilla.
Frontiers in plant science (2017-08-05)
Sarah Symanczik, Michelle Gisler, Cécile Thonar, Klaus Schlaeppi, Marcel Van der Heijden, Ansgar Kahmen, Thomas Boller, Paul Mäder
ABSTRACT
Naranjilla (
Sigma-Aldrich
Benzenesulfonyl hydrazide, 98%
Pricing and availability is not currently available.