Merck
  • New reporter gene assays for detecting natural and synthetic molting hormone agonists using yeasts expressing ecdysone receptors of various insects.
New reporter gene assays for detecting natural and synthetic molting hormone agonists using yeasts expressing ecdysone receptors of various insects.
FEBS open bio (2017-07-07)
Sayoko Ito-Harashima, Mai Matsuura, Masanobu Kawanishi, Yoshiaki Nakagawa, Takashi Yagi
ABSTRACT
Synthetic nonsteroidal ecdysone agonists, a class of insect growth regulators (IGRs), target the ecdysone receptor (EcR), which forms a heterodimer with ultraspiracle (USP) to transactivate ecdysone response genes. These compounds have high binding affinities to the EcR-USP complexes of certain insects and their toxicity is selective for certain taxonomic orders. In the present study, we developed reporter gene assay (RGA) systems to detect molting hormone (ecdysone) activity by introducing EcR-USP cDNA and a bacterial
Product Description
Supelco
Tebufenozide, PESTANAL®, analytical standard
Sigma-Aldrich
20-Hydroxyecdysone, ≥93% (HPLC), powder
Sigma-Aldrich
Ponasterone A, ≥65%
Sigma-Aldrich
Aldosterone, ≥95% (HPLC)
Supelco
Halofenozide, PESTANAL®, analytical standard
Supelco
Methoxyfenozide, PESTANAL®, analytical standard