Merck
  • Detection and quantification of 12 anabolic steroids and analogs in human whole blood and 20 in hair using LC-HRMS/MS: application to real cases.
Detection and quantification of 12 anabolic steroids and analogs in human whole blood and 20 in hair using LC-HRMS/MS: application to real cases.
International journal of legal medicine (2017-02-27)
Nicolas Fabresse, Stanislas Grassin-Delyle, Isabelle Etting, Jean-Claude Alvarez
ABSTRACT
We developed and validated a method to detect and quantify 12 anabolic steroids in blood (androstenedione, dihydrotestosterone, boldenone, epitestosterone, mesterolone, methandienone, nandrolone, stanozolol, norandrostenedione, tamoxifene, testosterone, trenbolone) and eight more in hair samples (nandrolone phenylpropionate, nandrolone decanoate, testosterone propionate, testosterone benzoate, testosterone cypionate, testosterone decanoate, testosterone phenylpropionate, testosterone undecanoate) using liquid chromatography coupled to high-resolution mass spectrometry. This method used a benchtop Orbitrap mass spectrometer operating with an APCI probe under positive ionization mode. Analysis was realized in full scan experiment with a nominal resolving power of 140,000. After addition of the internal standard (testosterone-D3) and incubation in phosphate buffer pH = 5 for hair, 200 μL of blood and 30 mg of hair samples were extracted with heptane. LOQ and LOD were determined at 5 and 1 ng mL
Product Description
Supelco
4-Androstene-3,17-dione, VETRANAL®, analytical standard
Sigma-Aldrich
Phenyl propionate, AldrichCPR