Merck
  • A novel method for detection and ex vivo expansion of HIV type 1-specific cytolytic T lymphocytes.
A novel method for detection and ex vivo expansion of HIV type 1-specific cytolytic T lymphocytes.
AIDS research and human retroviruses (1994-11-01)
M N Lubaki, M A Egan, R F Siliciano, K J Weinhold, R C Bollinger
ABSTRACT
Studies have shown that cytolytic T lymphocyte (CTL) responses may be critical to the clearance of the early viremia in acute HIV-1 infection. It is likely that these cells play an important role in prolonging the asymptomatic phase of the infection. Although HIV-1-specific CTL activity can be detected in direct assays of freshly isolated peripheral blood lymphocytes (PBL) from some infected individuals, this method fails to detect CTL that are present at low frequency and resting, memory CTL. For these reasons, direct CTL assays on PBL from seropositive individuals may underestimate the level of CTL immunity. As part of ongoing investigations of CTL activity in HIV-1-infected individuals, we developed a novel strategy for the detection and ex vivo expansion of HIV-1-specific CTL. This technique involves selective stimulation of PBL from seropositive individuals with autologous Epstein-Barr virus (EBV)-transformed, B-lymphoblastoid cell lines (B-LCL) infected with vaccinia vectors expressing various HIV-1 genes. Prior to their use for in vitro stimulation, B-LCL are treated with psoralen and UV light to inactivate vaccinia virus. After 1 week of stimulation, CTL activity in stimulated cultures is measured in a standard 51Cr release assay. This ex vivo expansion method can selectively increase the bulk culture CTL activity against env, gag and nef, even in some seropositive individuals with low CD4 counts and little evidence of HIV-1-specific CTL in assays of freshly isolated PBL. These expanded CTL are predominantly of the CD8+ phenotype.(ABSTRACT TRUNCATED AT 250 WORDS)
Sigma-Aldrich
4′-Aminomethyltrioxsalen hydrochloride
Pricing and availability is not currently available.