Merck
  • Development and characterization of embelin-loaded nanolipid carriers for brain targeting.
Development and characterization of embelin-loaded nanolipid carriers for brain targeting.
Artificial cells, nanomedicine, and biotechnology (2016-03-26)
Nancy Sharma, Saurav Bhandari, Rahul Deshmukh, Awesh K Yadav, Neeraj Mishra
ABSTRACT
The objective of present work was to enquire the potential use of embelin-loaded nanolipid carriers for brain targeting. The average particle size and polydispersity index (PDI) of optimized formulation (F19) were found to be 152 ± 19.7 nm and 0.143 ± 0.023, respectively. Nanolipid carrier (NLC) was also significantly attenuated pentylenetetrazole (PTZ)-induced biochemical parameters in comparison to plain embelin that results in an increase in the level of malondialdehyde (MDA), nitrite, and reduction in the level of glutathione. From the results, it was concluded that embelin-NLCs developed as a beneficial carrier to achieve sustained release and brain targeting through nasal route.
Sigma-Aldrich
2-Octyl-1-dodecanol, 97%
Pricing and availability is not currently available.