Merck
  • A sensitive voltammetric detection of pramipexole based on 1,1,3,3-tetramethyldisilazanecarbon nanotube modified electrode.
A sensitive voltammetric detection of pramipexole based on 1,1,3,3-tetramethyldisilazanecarbon nanotube modified electrode.
Materials science & engineering. C, Materials for biological applications (2017-04-19)
Saeede Amirighadi, Jahan Bakhsh Raoof, Fereshteh Chekin, Reza Ojani
ABSTRACT
A simple low-cost method is proposed to fabricate a functionalized multi-wall carbon nanotube (MWCNT) with 1,1,3,3-tetramethyldisilazane (TMDS) molecule. The techniques of scanning electron microscope (SEM) with electron diffraction and energy dispersive X-ray (EDAX) analysis were applied to characterize the functionalized TMDS-MWCNT. The results showed that a MWCNT with high functionalization of TMDS can be obtained using this simple method. A new nanostructure sensor was constructed based on a glassy carbon electrode modified with TMDS-MWCNT (TMDS-MWCNT/GCE). It was found that the TMDS-MWCNT/GCE exhibits good catalytic activity toward oxidation of pramipexole (PPX) drug, leading to a concentration range of 0.8 to 600μM with a detection limit of 0.2μM at 3σ using the differential pulse voltammetry technique, a sensitivity of 0.084μAμM
Sigma-Aldrich
1,1,3,3-Tetramethyldisilazane, 97%