Merck
  • Spatiotemporal distribution and potential sources of perfluoroalkyl acids in Huangpu River, Shanghai, China.
Spatiotemporal distribution and potential sources of perfluoroalkyl acids in Huangpu River, Shanghai, China.
Chemosphere (2017-02-06)
Zhuyu Sun, Chaojie Zhang, Hong Yan, Changlai Han, Ling Chen, Xiangzhou Meng, Qi Zhou
ABSTRACT
Perfluoroalkyl acids (PFAAs) have been found to be ubiquitously disseminated in the environment due to their widespread use in recent decades. In this study, the occurrence and spatiotemporal distribution of PFAAs in the surface water of Huangpu River, Shanghai, China were investigated from 2012 to 2014. The total concentration of 14 PFAAs (ΣPFAAs) ranged from 39.8 to 596.2 ng L
Sigma-Aldrich
Undecafluorohexanoic acid, ≥97.0% (T)
Sigma-Aldrich
Pentafluoropropionic acid, 97%
Sigma-Aldrich
Tridecafluorohexane-1-sulfonic acid potassium salt, ≥98.0% (T)
Sigma-Aldrich
Perfluoroheptanoic acid, ≥97.0%
Sigma-Aldrich
Heptadecafluorooctanesulfonic acid solution, ~40% in H2O (T)
Sigma-Aldrich
Perfluoropentanoic acid, 97%
Sigma-Aldrich
Perfluoroundecanoic acid, 95%
Sigma-Aldrich
Perfluorononanoic acid, 97%
Sigma-Aldrich
Perfluorotetradecanoic acid, 96%
Sigma-Aldrich
Tricosafluorododecanoic acid, 95%
Sigma-Aldrich
Perfluorodecanoic acid, 98%