Merck
  • Interaction between RNF8 and DYRK2 is required for the recruitment of DNA repair molecules to DNA double-strand breaks.
Interaction between RNF8 and DYRK2 is required for the recruitment of DNA repair molecules to DNA double-strand breaks.
FEBS letters (2017-02-15)
Takenori Yamamoto, Naoe Taira Nihira, Satomi Yogosawa, Katsuhiko Aoki, Hiroyuki Takeda, Tatsuya Sawasaki, Kiyotsugu Yoshida
ABSTRACT
The genome of eukaryotic cells is frequently exposed to damage by various genotoxins. Phosphorylation of histone H2AX at Serine 139 (γ-H2AX) is a hallmark of DNA damage. RNF8 monoubiquitinates γ-H2AX with the Lys63-linked ubiquitin chain to tether DNA repair molecules at DNA lesions. A high-throughput screening identified RNF8 as a binding partner of dual-specificity tyrosine phosphorylation-regulated kinase 2 (DYRK2). Notably, DNA damage-induced monoubiquitination of γ-H2AX is impaired in DYRK2-depleted cells. The foci formation of p53-binding protein 1 at DNA double-strand break sites is suppressed in DYRK2 knockdown cells, which fail to repair the DNA damage. A homologous recombination assay showed decreased repair efficiency in DYRK2-depleted cells. Our findings indicate direct interaction of DYRK2 with RNF8 in regulating response to DNA damage.
Sigma-Aldrich
7-Aminoactinomycin D, ~97% (HPLC), powder
Sigma-Aldrich
MISSION® esiRNA, targeting human DYRK2