Merck
  • Microalgae cultivation in urban wastewater: Coelastrum cf. pseudomicroporum as a novel carotenoid source and a potential microalgae harvesting tool.
Microalgae cultivation in urban wastewater: Coelastrum cf. pseudomicroporum as a novel carotenoid source and a potential microalgae harvesting tool.
Bioresource technology (2017-01-09)
Bárbara Úbeda, José Ángel Gálvez, Mónica Michel, Ana Bartual
ABSTRACT
The aim of this work was to study the optimal growth and high value-added production of the microalgae Coelastrum cf. pseudomicroporum Korshikov cultivated in urban wastewater. It was observed that C. cf. pseudomicroporum grew ideally in this medium, acting as an efficient nutrient starver. Additionally, the obtained biomass increased carotenoid cell content after saltwater stress. The effects of light intensity and salt stress on its growth rate were analysed. The results showed that this alga can grow very fast using wastewater as culture medium, reaching maximum growth rates of 1.61±0.05day
Sigma-Aldrich
trans,trans-2,4-Heptandienal, ≥88%
Sigma-Aldrich
trans,trans-2,4-Decadienal, technical grade, 85%