Merck
  • Disulfiram moderately restores impaired hepatic redox status of rats subchronically exposed to cadmium.
Disulfiram moderately restores impaired hepatic redox status of rats subchronically exposed to cadmium.
Journal of enzyme inhibition and medicinal chemistry (2017-01-20)
Aida Begic, Ana Djuric, Milica Ninkovic, Ivana Stevanovic, Dragan Djurdjevic, Milos Pavlovic, Katarina Jelic, Ana Pantelic, Goran Zebic, Bratislav Dejanovic, Ivan Stanojevic, Danilo Vojvodic, Petar Milosavljevic, Mirjana Djukic, Luciano Saso
ABSTRACT
Examination of cadmium (Cd) toxicity and disulfiram (DSF) effect on liver was focused on oxidative stress (OS), bioelements status, morphological and functional changes. Male Wistar rats were intraperitoneally treated with 1 mg CdCl
Sigma-Aldrich
Glutathione-S-Transferase (GST) Assay Kit, sufficient for 500 multiwell tests
Sigma-Aldrich
Glutathione Reductase Assay Kit, Sufficient for 100 colorimetric tests
Sigma-Aldrich
Glutathione Peroxidase Cellular Activity Assay Kit, Sufficient for 100 colorimetric tests