Merck
  • Influence of Pre-etching Times on Fatigue Strength of Self-etch Adhesives to Enamel.
Influence of Pre-etching Times on Fatigue Strength of Self-etch Adhesives to Enamel.
The journal of adhesive dentistry (2016-12-10)
Toshiki Takamizawa, Wayne W Barkmeier, Akimasa Tsujimoto, Hajime Endo, Kenji Tsuchiya, Robert L Erickson, Mark A Latta, Masashi Miyazaki
ABSTRACT
To use shear bond strength (SBS) and shear fatigue strength (SFS) testing to determine the influence of phosphoric acid pre-etching times prior to application of self-etch adhesives on enamel bonding. Two single-step self-etch universal adhesives (Prime&Bond Elect and Scotchbond Universal), a conventional single-step self-etch adhesive (G-ӕnial Bond), and a conventional two-step self-etch adhesive (OptiBond XTR) were used. The SBS and SFS were obtained with phosphoric acid pre-etching for 3, 10, or 15 s prior to application of the adhesives, and without pre-etching (0 s) as a control. A staircase method was used to determine the SFS with 10 Hz frequency for 50,000 cycles or until failure occurred. The mean demineralization depth for each treated enamel surface was also measured using a profilometer. For all the adhesives, the groups with pre-etching showed significantly higher SBS and SFS than groups without pre-etching. However, there was no significant difference in SBS and SFS among groups with > 3 s of preetching. In addition, although the groups with pre-etching showed significantly deeper demineralization depths than groups without pre-etching, there was no significant difference in depth among groups with > 3 s of pre-etching. Three seconds of phosphoric acid pre-etching prior to application of self-etch adhesive can enhance enamel bonding effectiveness.
Sigma-Aldrich
Dipentaerythritol, technical grade