Merck
  • Hyperventilation assists proarrhythmia development during delayed repolarization in clofilium-treated, anaesthetized, mechanically ventilated rabbits.
Hyperventilation assists proarrhythmia development during delayed repolarization in clofilium-treated, anaesthetized, mechanically ventilated rabbits.
Journal of physiology and pharmacology : an official journal of the Polish Physiological Society (2016-12-25)
H Papp, A Sarusi, A S Farkas, H Takacs, P Kui, D Vincze, E Ivany, A Varro, J G Papp, T Forster, A Farkas
ABSTRACT
Hyperventilation reduces partial pressure of CO
Sigma-Aldrich
Clofilium tosylate, >97%, solid