Merck
  • Profiling adrenal 11β-hydroxyandrostenedione metabolites in prostate cancer cells, tissue and plasma: UPC
Profiling adrenal 11β-hydroxyandrostenedione metabolites in prostate cancer cells, tissue and plasma: UPC
The Journal of steroid biochemistry and molecular biology (2016-06-28)
Therina du Toit, Liezl M Bloem, Jonathan L Quanson, Riaan Ehlers, Antonio M Serafin, Amanda C Swart
ABSTRACT
Adrenal C
Sigma-Aldrich
Cortisol-9,11,12,12-d4, ≥98 atom % D, ≥98% (CP)