- Synthesis and catalytic properties of silver nanoparticles supported on porous cellulose acetate sheets and wet-spun fibers.
Synthesis and catalytic properties of silver nanoparticles supported on porous cellulose acetate sheets and wet-spun fibers.
Carbohydrate polymers (2016-12-19)
Tahseen Kamal, Ikram Ahmad, Sher Bahadar Khan, Abdullah M Asiri
PMID27987930
ABSTRACT
Cellulose acetate fibers (CA