Merck
  • Cell Entry of Porcine Epidemic Diarrhea Coronavirus Is Activated by Lysosomal Proteases.
Cell Entry of Porcine Epidemic Diarrhea Coronavirus Is Activated by Lysosomal Proteases.
The Journal of biological chemistry (2016-10-13)
Chang Liu, Yuanmei Ma, Yang Yang, Yuan Zheng, Jian Shang, Yusen Zhou, Shibo Jiang, Lanying Du, Jianrong Li, Fang Li
ABSTRACT
Porcine epidemic diarrhea coronavirus (PEDV) is currently devastating the United States pork industry by causing an 80-100% fatality rate in infected piglets. Coronavirus spike proteins mediate virus entry into cells, a process that requires the spike proteins to be proteolytically activated. It has been a conundrum which proteases activate PEDV entry. Here we systematically investigated the roles of different proteases in PEDV entry using pseudovirus entry, biochemical, and live virus infection assays. We found that the PEDV spike is activated by lysosomal cysteine proteases but not proprotein convertases or cell surface serine proteases. Extracellular trypsin activates PEDV entry when lysosomal cysteine proteases are inhibited. We further pinpointed cathepsin L and cathepsin B as the lysosomal cysteine proteases that activate the PEDV spike. These results advance our understanding of the molecular mechanism for PEDV entry and identify potential antiviral targets for curbing the spread of PEDV.
Sigma-Aldrich
Camostat mesylate, ≥98% (HPLC)
Sigma-Aldrich
E-64d, protease inhibitor