Merck
  • Carbon emissions, logistics volume and GDP in China: empirical analysis based on panel data model.
Carbon emissions, logistics volume and GDP in China: empirical analysis based on panel data model.
Environmental science and pollution research international (2016-09-24)
Xiaopeng Guo, Dongfang Ren, Jiaxing Shi
ABSTRACT
This paper studies the relationship among carbon emissions, GDP, and logistics by using a panel data model and a combination of statistics and econometrics theory. The model is based on the historical data of 10 typical provinces and cities in China during 2005-2014. The model in this paper adds the variability of logistics on the basis of previous studies, and this variable is replaced by the freight turnover of the provinces. Carbon emissions are calculated by using the annual consumption of coal, oil, and natural gas. GDP is the gross domestic product. The results showed that the amount of logistics and GDP have a contribution to carbon emissions and the long-term relationships are different between different cities in China, mainly influenced by the difference among development mode, economic structure, and level of logistic development. After the testing of panel model setting, this paper established a variable coefficient model of the panel. The influence of GDP and logistics on carbon emissions is obtained according to the influence factors among the variables. The paper concludes with main findings and provides recommendations toward rational planning of urban sustainable development and environmental protection for China.
Sigma-Aldrich
Zinc isopropylsulfinate, 95%