Merck
  • Pias3-dependent SUMOylation controls mammalian cone photoreceptor differentiation.
Pias3-dependent SUMOylation controls mammalian cone photoreceptor differentiation.
Nature neuroscience (2010-08-24)
Akishi Onishi, Guang-Hua Peng, Shiming Chen, Seth Blackshaw
ABSTRACT
Selective expression of retinal cone opsin genes is essential for color vision, but the mechanism mediating this process is poorly understood. Both vertebrate rod and medium wavelength-sensitive (M) cone photoreceptors differentiate by repression of a short wavelength-sensitive (S) cone differentiation program. We found that Pias3 acts in mouse cone photoreceptors to activate expression of M opsin and repress expression of S opsin, with the transcription factors Trbeta2 and Rxrgamma mediating preferential expression of Pias3 in M cones. Finally, we observed that Pias3 directly regulated M and S cone opsin expression by modulating the cone-enriched transcription factors Rxrgamma, Roralpha and Trbeta1. Our results indicate that Pias3-dependent SUMOylation of photoreceptor-specific transcription factors is a common mechanism that controls both rod and cone photoreceptor subtype specification, regulating distinct molecular targets in the two cell types.
Sigma-Aldrich
Monoclonal Anti-PIAS3 antibody produced in mouse, ~2 mg/mL, clone PIA3, purified immunoglobulin, buffered aqueous solution