Merck
  • Matrix Metalloproteinase 10 Degradomics in Keratinocytes and Epidermal Tissue Identifies Bioactive Substrates With Pleiotropic Functions.
Matrix Metalloproteinase 10 Degradomics in Keratinocytes and Epidermal Tissue Identifies Bioactive Substrates With Pleiotropic Functions.
Molecular & cellular proteomics : MCP (2015-10-18)
Pascal Schlage, Tobias Kockmann, Fabio Sabino, Jayachandran N Kizhakkedathu, Ulrich Auf dem Keller
ABSTRACT
Matrix metalloproteinases (MMPs) are important players in skin homeostasis, wound repair, and in the pathogenesis of skin cancer. It is now well established that most of their functions are related to processing of bioactive proteins rather than components of the extracellular matrix (ECM). MMP10 is highly expressed in keratinocytes at the wound edge and at the invasive front of tumors, but hardly any non-ECM substrates have been identified and its function in tissue repair and carcinogenesis is unclear. To better understand the role of MMP10 in the epidermis, we employed multiplexed iTRAQ-based Terminal Amine Isotopic Labeling of Substrates (TAILS) and monitored MMP10-dependent proteolysis over time in secretomes from keratinocytes. Time-resolved abundance clustering of neo-N termini classified MMP10-dependent cleavage events by efficiency and refined the MMP10 cleavage site specificity by revealing a so far unknown preference for glutamate in the P1 position. Moreover, we identified and validated the integrin alpha 6 subunit, cysteine-rich angiogenic inducer 61 and dermokine as novel direct MMP10 substrates and provide evidence for MMP10-dependent but indirect processing of phosphatidylethanolamine-binding protein 1. Finally, we sampled the epidermal proteome and degradome in unprecedented depth and confirmed MMP10-dependent processing of dermokine in vivo by TAILS analysis of epidermis from transgenic mice that overexpress a constitutively active mutant of MMP10 in basal keratinocytes. The newly identified substrates are involved in cell adhesion, migration, proliferation, and/or differentiation, indicating a contribution of MMP10 to local modulation of these processes during wound healing and cancer development. Data are available via ProteomeXchange with identifier PXD002474.
Sigma-Aldrich
Anti-ITGA6 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-PEBP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-CYR61 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution