Merck
  • Pigment Epithelium-derived Factor (PEDF) Blocks Wnt3a Protein-induced Autophagy in Pancreatic Intraepithelial Neoplasms.
Pigment Epithelium-derived Factor (PEDF) Blocks Wnt3a Protein-induced Autophagy in Pancreatic Intraepithelial Neoplasms.
The Journal of biological chemistry (2016-08-26)
Jingjing Gong, Glenn Belinsky, Usman Sagheer, Xuchen Zhang, Paul J Grippo, Chuhan Chung
ABSTRACT
An increase in autophagy characterizes pancreatic carcinogenesis, but the signals that regulate this process are incompletely understood. Because canonical Wnt/β-catenin signaling is necessary for the transition from early to advanced pancreatic intraepithelial neoplasia (PanIN) lesions, we assessed whether Wnt ligands and endogenous inhibitors of Wnt signaling modulate autophagy. In this study, canonical Wnt3a ligand induced autophagy markers and vacuoles in murine PanIN cells. Furthermore, pigment epithelium-derived factor (PEDF), a secreted glycoprotein known for its anti-tumor properties, blocked Wnt3a-directed induction of autophagy proteins. Autophagy inhibition was complemented by reciprocal regulation of the oxidative stress enzymes, superoxide dismutase 2 (SOD2) and catalase. Transcriptional control of Sod2 expression was mediated by PEDF-induced NFκB nuclear translocation. PEDF-dependent SOD2 expression in PanIN lesions was recapitulated in a murine model of PanIN formation where PEDF was deleted. In human PanIN lesions, co-expression of PEDF and SOD2 was observed in the majority of early PanIN lesions (47/50, 94%), whereas PEDF and SOD2 immunolocalization in high-grade human PanIN-2/3 was uncommon (7/50, 14%). These results indicate that PEDF regulates autophagy through coordinate Wnt signaling blockade and NFκB activation.
Sigma-Aldrich
Anti-Pigment Epithelium Derived Factor Antibody, clone 10F12.2, clone 10F12.2, Chemicon®, from mouse