Merck
  • Stem Cells Antigen-1 Enriches for a Cancer Stem Cell-Like Subpopulation in Mouse Gastric Cancer.
Stem Cells Antigen-1 Enriches for a Cancer Stem Cell-Like Subpopulation in Mouse Gastric Cancer.
Stem cells (Dayton, Ohio) (2016-02-13)
Jun Won Park, Jung Min Park, Dong Min Park, Dae-Yong Kim, Hark Kyun Kim
ABSTRACT
There is a strong need to identify markers to enrich gastric cancer stem cells (CSCs). However, CSC enrichment markers for mouse gastric cancers have not yet been determined. In our previous study, we generated primary mouse gastric cancer cell line NCC-S1 (S1) established from a Villin-cre;Smad4(F/F) ;Trp53(F/F) ;Cdh1(F/wt) mouse and its metastatic variant cell line NCC-S1M (S1M). Interestingly, S1M cells exhibited CSC-like features, such as increased tumorigenic potential and chemoresistance. By comparing gene expression profiles between S1 and S1M cells, we identified Stem Cells Antigen-1 (Sca-1) as a cell surface marker, which was mostly upregulated in S1M. Sca-1 was upregulated in tumorspheres from S1 cells or after cisplatin treatment in S1 cells. Immunofluorescence (IF) analysis showed that approximately 7% of cancer cells exhibited positivity for Sca-1 in primary mouse gastric cancer tissues. An in vivo-limiting dilution assay showed that Sca-1(high) mouse gastric cancer cells demonstrated increased tumorigenicity compared with Sca-1(negative) cells. The Sca-1 expression was downregulated by TGF-β pathway activation and Wnt pathway inhibition in mouse gastric cancer cells. Sca-1(high) cells showed relatively low TGF-β reporter activity and high TCF/LEF1 reporter activity compared with Sca-1(negative) cells. A chromatin immunoprecipitation analysis demonstrated that Sca-1 was a β-catenin/LEF1 target gene. Sca-1(high) allografts were more resistant to cisplatin/fluorouracil chemotherapy than Sca-1(negative) allografts, and overexpressed Bcl-xL. Eighty-five mouse genes overexpressed in Sca-1(high) S1 cells compared with Sca-1(negative) cells clustered 123 pretreatment gastric cancer patient samples according to survival following chemotherapy. Taken together, Sca-1 is a novel CSC enrichment marker that mediates TGF-β and Wnt/β-catenin signaling in mouse gastric cancer. Stem Cells 2016;34:1177-1187.
Sigma-Aldrich
FragEL DNA Fragmentation Detection Kit, Fluorescent - TdT Enzyme
Sigma-Aldrich
ChIPAb+ LEF1 - ChIP Validated Antibody and Primer Set, from mouse
Sigma-Aldrich
Lithium chloride, ≥99%