Merck
  • Breathable waveguides for combined light and CO2 delivery to microalgae.
Breathable waveguides for combined light and CO2 delivery to microalgae.
Bioresource technology (2016-03-22)
Scott C Pierobon, Jason Riordon, Brian Nguyen, David Sinton
ABSTRACT
Suboptimal light and chemical distribution (CO2, O2) in photobioreactors hinder phototrophic microalgal productivity and prevent economically scalable production of biofuels and bioproducts. Current strategies that improve illumination in reactors negatively impact chemical distribution, and vice versa. In this work, an integrated illumination and aeration approach is demonstrated using a gas-permeable planar waveguide that enables combined light and chemical distribution. An optically transparent cellulose acetate butyrate (CAB) slab is used to supply both light and CO2 at various source concentrations to cyanobacteria. The breathable waveguide architecture is capable of cultivating microalgae with over double the growth as achieved with impermeable waveguides.
Sigma-Aldrich
Cyanobacteria BG-11 Freshwater Solution, 50 ×, liquid, suitable for plant cell culture
Pricing and availability is not currently available.