Merck
Knockout of frataxin gene causes embryo lethality in Arabidopsis.
FEBS letters (2007-01-30)
Valentina Vazzola, Alessia Losa, Carlo Soave, Irene Murgia
ABSTRACT
Frataxin is present in mitochondria of all eukaryotes as well as in the cytoplasm of bacteria. In humans, reduced expression of frataxin is associated with Friedreich's ataxia, a recessive inherited neurodegenerative and cardiac disorder leading to reduced life expectancy. Experimental evidences suggest that frataxin acts as an iron-chaperone protein, donating iron to the proteins involved in [Fe-S] cluster assembly and heme synthesis. It also possibly contributes to the process of iron detoxification and storage. The frataxin homolog from Arabidopsis thaliana (AtFH) is a single nuclear-encoded gene targeted to mitochondria and sharing 65% similarity with animal frataxin. In the present work, we show that the knocking out of AtFH gene causes arrest of Arabidopsis embryo development at the globular stage. Consistently with that, we also show by in situ hybridization that AtFH is expressed, in wt Arabidopsis plants, in ovule primordia as well as in embryos at various stages of development, suggesting a key role of plant frataxin during embryogenesis.
Sigma-Aldrich
Eosin Y, Dye content ~99 %
Pricing and availability is not currently available.
Roche
Anti-Digoxigenin-AP, Fab fragments, from sheep
Pricing and availability is not currently available.
Roche
DIG RNA Labeling Kit (SP6/T7), sufficient for 2 x 10 labeling reactions, kit of 1 (12 components), suitable for hybridization, suitable for Southern blotting
Pricing and availability is not currently available.
Roche
BCIP, solution, >95% (HPLC), pkg of 3 mL (150 mg)
Pricing and availability is not currently available.