Merck
  • Transcription sites are developmentally regulated during the asexual cycle of Plasmodium falciparum.
Transcription sites are developmentally regulated during the asexual cycle of Plasmodium falciparum.
PloS one (2013-02-15)
Carolina B Moraes, Thierry Dorval, Mónica Contreras-Dominguez, Fernando de M Dossin, Michael A E Hansen, Auguste Genovesio, Lucio H Freitas-Junior
ABSTRACT
Increasing evidence shows that the spatial organization of transcription is an important epigenetic factor in eukaryotic gene regulation. The malaria parasite Plasmodium falciparum shows a remarkably complex pattern of gene expression during the erythrocytic cycle, paradoxically contrasting with the relatively low number of putative transcription factors encoded by its genome. The spatial organization of nuclear subcompartments has been correlated with the regulation of virulence genes. Here, we investigate the nuclear architecture of transcription during the asexual cycle of malaria parasites. As in mammals, transcription is organized into discrete nucleoplasmic sites in P. falciparum, but in a strikingly lower number of foci. An automated analysis of 3D images shows that the number and intensity of transcription sites vary significantly between rings and trophozoites, although the nuclear volume remains constant. Transcription sites are spatially reorganized during the asexual cycle, with a higher proportion of foci located in the outermost nuclear region in rings, whereas in trophozoites, foci are evenly distributed throughout the nucleoplasm. As in higher eukaryotes, transcription sites are predominantly found in areas of low chromatin density. Immunofluorescence analysis shows that transcription sites form an exclusive nuclear compartment, different from the compartments defined by the silenced or active chromatin markers. In conclusion, these data suggest that transcription is spatially contained in discrete foci that are developmentally regulated during the asexual cycle of malaria parasites and located in areas of low chromatin density.
Sigma-Aldrich
Anti-acetyl-Histone H4 Antibody, 1 mg/mL, Upstate®
Sigma-Aldrich
L-α-Lysophosphatidylcholine from egg yolk, ≥99%, Type I, powder